4,4'-(1E,1'E)-2,2'-(1-Phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl)diphenol

Lieferant: Apollo Scientific

Synonyme: 4,4'-[(1-Phenylpyrazole-3,5-diyl)bis(ethene-2,1-diyl)]diphenol

OR471179-1G
APOSOR471179-1GEA 601 EUR
APOSOR471179-1G
4,4'-(1E,1'E)-2,2'-(1-Phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl)diphenol
4,4'-(1E,1'E)-2,2'-(1-Phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl)diphenol
Formel: C₂₅H₂₀N₂O₂
Molecular Weight: 380,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18433540
CAS-Nummer: 828911-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR