Ethyl 4-(Boc-aminomethyl)benzoate

Lieferant: Apollo Scientific

OR471182-5G
APOSOR471182-5GEA 601 EUR
APOSOR471182-5G
Ethyl 4-(Boc-aminomethyl)benzoate
Ethyl 4-(Boc-aminomethyl)benzoate
Formel: C₁₅H₂₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD10609786
CAS-Nummer: 157311-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR