6-(2-Chlorophenyl)morpholin-3-one
Lieferant: Apollo Scientific
OR471183-1G
APOSOR471183-1GEA
1010
EUR
APOSOR471183-1G
6-(2-Chlorophenyl)morpholin-3-one
6-(2-Chlorophenyl)morpholin-3-one
Formel:
C₁₀H₁₀ClNO₂ Molecular Weight: 211,645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09831998 CAS-Nummer: 951626-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...