1-(4-Chlorophenyl)pyrazole-4-carboxylic acid
Lieferant: Apollo Scientific
OR471186-1G
APOSOR471186-1GEA
662
EUR
APOSOR471186-1G
1-(4-Chlorophenyl)pyrazole-4-carboxylic acid
1-(4-Chlorophenyl)pyrazole-4-carboxylic acid
Formel:
C₁₀H₇ClN₂O₂ Molecular Weight: 222,628 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09936018 CAS-Nummer: 138907-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...