1-(3-Bromopropyl)-4-methylpiperazine dihydrobromide

Lieferant: Apollo Scientific

OR471187-1G
APOSOR471187-1GEA 397 EUR
APOSOR471187-1G
1-(3-Bromopropyl)-4-methylpiperazine dihydrobromide
1-(3-Bromopropyl)-4-methylpiperazine dihydrobromide
Formel: C₈H₁₇BrN₂·2BrH
Molecular Weight: 382,962 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03452742
CAS-Nummer: 5845-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR