4-(3-Bromopropyl)morpholine hydrobromide

Lieferant: Apollo Scientific

OR471189-1G OR471189-5G
APOSOR471189-1GEA 109 EUR
APOSOR471189-1G APOSOR471189-5G
4-(3-Bromopropyl)morpholine hydrobromide
4-(3-Bromopropyl)morpholine hydrobromide
Formel: C₇H₁₄BrNO·BrH
Molecular Weight: 289.008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176498
CAS-Nummer: 88806-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR