1-(3-Methoxypropyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

OR471190-1G
APOSOR471190-1GEA 916 EUR
APOSOR471190-1G
1-(3-Methoxypropyl)cyclopropanecarboxylic acid
1-(3-Methoxypropyl)cyclopropanecarboxylic acid
Formel: C₈H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD21669719
CAS-Nummer: 1468755-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR