1-(2-Methoxyethyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

OR471191-1G
APOSOR471191-1GEA 916 EUR
APOSOR471191-1G
1-(2-Methoxyethyl)cyclopropanecarboxylic acid
1-(2-Methoxyethyl)cyclopropanecarboxylic acid
Formel: C₇H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD21669750
CAS-Nummer: 1423031-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR