4-(4-Methoxyphenyl)tetrahydropyran-4-carboxylic acid

Lieferant: Apollo Scientific

OR471192-1G
APOSOR471192-1GEA 519 EUR
APOSOR471192-1G
4-(4-Methoxyphenyl)tetrahydropyran-4-carboxylic acid
4-(4-Methoxyphenyl)tetrahydropyran-4-carboxylic acid
Formel: C₁₃H₁₆O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04061780
CAS-Nummer: 3648-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR