Ethyl-2-chlorobenzoylformate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(2-chlorophenyl)-2-oxoacetate
OR471193-1G
APOSOR471193-1GEA
397
EUR
APOSOR471193-1G
Ethyl-2-chlorobenzoylformate
Ethyl-2-chlorobenzoylformate
Formel:
C₁₀H₉ClO₃ Molecular Weight: 212,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801392 CAS-Nummer: 62123-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...