4-Bromo-1-methylpiperidine hydrobromide

Lieferant: Apollo Scientific

OR471196-1G
APOSOR471196-1GEA 713 EUR
APOSOR471196-1G
4-Bromo-1-methylpiperidine hydrobromide
4-Bromo-1-methylpiperidine hydrobromide
Formel: C₆H₁₂BrN·BrH
Molecular Weight: 258,982 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218726
CAS-Nummer: 54288-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR