3-(2-Chlorophenyl)cyclobutanone

Lieferant: Apollo Scientific

OR471197-1G
APOSOR471197-1GEA 0 EUR
APOSOR471197-1G
3-(2-Chlorophenyl)cyclobutanone
3-(2-Chlorophenyl)cyclobutanone
Formel: C₁₀H₉ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD11848525
CAS-Nummer: 1080636-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR