1-Acetyl-3-aminopyrrole

Lieferant: Apollo Scientific

OR471198-1G
APOSOR471198-1GEA 601 EUR
APOSOR471198-1G
1-Acetyl-3-aminopyrrole
1-Acetyl-3-aminopyrrole
Formel: C₆H₈N₂O
Molecular Weight: 124,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28385453
CAS-Nummer: 1823438-94-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR