4-Cbz-piperazine-1-carbonyl chloride
Lieferant: Apollo Scientific
OR471201-1G
APOSOR471201-1GEA
448
EUR
APOSOR471201-1G
4-Cbz-piperazine-1-carbonyl chloride
4-Cbz-piperazine-1-carbonyl chloride
Formel:
C₁₃H₁₅ClN₂O₃ Molecular Weight: 282,723 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743772 CAS-Nummer: 25539-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...