4-Isopropylpiperazine-1-carbonyl chloride
Lieferant: Apollo Scientific
OR471202-1G
APOSOR471202-1GEA
448
EUR
APOSOR471202-1G
4-Isopropylpiperazine-1-carbonyl chloride
4-Isopropylpiperazine-1-carbonyl chloride
Formel:
C₈H₁₅ClN₂O Molecular Weight: 190,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13174146 CAS-Nummer: 745731-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...