Methyl 2-Amidinoacetate hydrochloride

Lieferant: Apollo Scientific
Achtung

OR471204-1G
APOSOR471204-1GEA 601 EUR
APOSOR471204-1G
Methyl 2-Amidinoacetate hydrochloride
Methyl 2-Amidinoacetate hydrochloride
Formel: C₄H₈N₂O₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22070092
CAS-Nummer: 103173-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR