3-(Cyclopropylamino)propionic acid

Lieferant: Apollo Scientific

Synonyme: 3-(Cyclopropylamino)propionic Acid

OR471206-1G
APOSOR471206-1GEA 405 EUR
APOSOR471206-1G
3-(Cyclopropylamino)propionic acid
3-(Cyclopropylamino)propionic acid
Formel: C₆H₁₁NO₂
Molecular Weight: 129.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208357
CAS-Nummer: 202059-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR