Ethyl-3-(chlorosulphonyl)propanoate

Lieferant: Apollo Scientific
Gefahr

OR471207-1G
APOSOR471207-1GEA 438 EUR
APOSOR471207-1G
Ethyl-3-(chlorosulphonyl)propanoate
Ethyl-3-(chlorosulphonyl)propanoate
Formel: C₅H₉ClO₄S
Molecular Weight: 200,64 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07186267
CAS-Nummer: 103472-25-9
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR