Ethyl 2-Oxo-2-[(2-oxo-2-phenylethyl)amino]acetate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-oxo-2-((2-oxo-2-phenylethyl)amino)acetate

OR471209-1G
APOSOR471209-1GEA 493 EUR
APOSOR471209-1G
Ethyl 2-Oxo-2-[(2-oxo-2-phenylethyl)amino]acetate
Ethyl 2-Oxo-2-[(2-oxo-2-phenylethyl)amino]acetate
Formel: C₁₂H₁₃NO₄
Molecular Weight: 235,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22493454
CAS-Nummer: 84978-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR