8-Iodoquinazolin-4(3H)-one

Lieferant: Apollo Scientific

OR471210-1G
APOSOR471210-1GEA 119 EUR
APOSOR471210-1G
8-Iodoquinazolin-4(3H)-one
8-Iodoquinazolin-4(3H)-one
Formel: C₈H₅IN₂O
Molecular Weight: 272,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954852
CAS-Nummer: 77150-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR