Diethyl 2-(4-Chlorobenzamido)malonate

Lieferant: Apollo Scientific

OR471211-1G
APOSOR471211-1GEA 601 EUR
APOSOR471211-1G
Diethyl 2-(4-Chlorobenzamido)malonate
Diethyl 2-(4-Chlorobenzamido)malonate
Formel: C₁₄H₁₆ClNO₅
Molecular Weight: 313,734 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09031363
CAS-Nummer: 81918-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR