3-Amino-2-(aminomethyl)pyridine

Lieferant: Apollo Scientific

OR471262-1G
APOSOR471262-1GEA 1120 EUR
APOSOR471262-1G
3-Amino-2-(aminomethyl)pyridine
3-Amino-2-(aminomethyl)pyridine
Formel: C₆H₉N₃
Molecular Weight: 123,156 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08703173
CAS-Nummer: 144288-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR