5-Amino-4-(aminomethyl)-1-methylpyrazole

Lieferant: Apollo Scientific

OR471263-1G
APOSOR471263-1GEA 1010 EUR
APOSOR471263-1G
5-Amino-4-(aminomethyl)-1-methylpyrazole
5-Amino-4-(aminomethyl)-1-methylpyrazole
Formel: C₅H₁₀N₄
Molecular Weight: 126,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213315
CAS-Nummer: 753444-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR