4-(Aminomethyl)-5-bromo-2-(methylthio)pyrimidine

Lieferant: Apollo Scientific

OR471265-1G
APOSOR471265-1GEA 916 EUR
APOSOR471265-1G
4-(Aminomethyl)-5-bromo-2-(methylthio)pyrimidine
4-(Aminomethyl)-5-bromo-2-(methylthio)pyrimidine
Formel: C₆H₈BrN₃S
Molecular Weight: 234,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28347180
CAS-Nummer: 1823402-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR