2-(Aminomethyl)-3,5-dimethylpyrrole

Lieferant: Apollo Scientific

OR471267-1G
APOSOR471267-1GEA 1120 EUR
APOSOR471267-1G
2-(Aminomethyl)-3,5-dimethylpyrrole
2-(Aminomethyl)-3,5-dimethylpyrrole
Formel: C₇H₁₂N₂
Molecular Weight: 124.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28398135
CAS-Nummer: 1503251-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR