5-(Aminomethyl)-1,4-benzodioxane
Lieferant: Apollo Scientific
OR471269-5G
APOSOR471269-5GEA
1540
EUR
APOSOR471269-5G
5-(Aminomethyl)-1,4-benzodioxane
5-(Aminomethyl)-1,4-benzodioxane
Formel:
C₉H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972618 CAS-Nummer: 261633-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...