1H-Indol-6-ylmethylamine 96%

Lieferant: Apollo Scientific

Synonyme: (1H-Indol-6-yl)methanamine, 6-(Aminomethyl)indole

OR471270-5G OR471270-1G OR471270-25G
APOSOR471270-5GEA 319 EUR
APOSOR471270-5G APOSOR471270-1G APOSOR471270-25G
1H-Indol-6-ylmethylamine 96%
1H-Indol-6-ylmethylamine
Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213839
CAS-Nummer: 3468-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR