3-Phenylbutylamine

Lieferant: Apollo Scientific

OR471278-1G
APOSOR471278-1GEA 418 EUR
APOSOR471278-1G
3-Phenylbutylamine
3-Phenylbutylamine
Formel: C₁₀H₁₅N
Molecular Weight: 149,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07786703
CAS-Nummer: 38135-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR