4-(2,6-Dichlor-4-pyrimidinyl)morpholin
Lieferant: Apollo Scientific
Synonyme:
4-(2,6-Dichloropyrimidin-4-yl)morpholine, 2,4-Dichloro-6-(4-morpholino)pyrimidine
OR471279-5G
APOSOR471279-5GEA
366
EUR
APOSOR471279-5G
4-(2,6-Dichlor-4-pyrimidinyl)morpholin
4-(2,6-Dichlor-4-pyrimidinyl)morpholin
Formel:
C₈H₉Cl₂N₃O Molecular Weight: 234,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09746259 CAS-Nummer: 52127-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...