2-Methylthieno[2,3-d]pyrimidin-4(3H)-one

Lieferant: Apollo Scientific

OR471281-1G
APOSOR471281-1GEA 1120 EUR
APOSOR471281-1G
2-Methylthieno[2,3-d]pyrimidin-4(3H)-one
2-Methylthieno[2,3-d]pyrimidin-4(3H)-one
Formel: C₇H₆N₂OS
Molecular Weight: 166,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00706457
CAS-Nummer: 21582-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR