2,6-Dichloroquinazolin-4(3H)-one

Lieferant: Apollo Scientific

OR471282-5G
APOSOR471282-5GEA 1450 EUR
APOSOR471282-5G
2,6-Dichloroquinazolin-4(3H)-one
2,6-Dichloroquinazolin-4(3H)-one
Formel: C₈H₄Cl₂N₂O
Molecular Weight: 215,036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520689
CAS-Nummer: 20197-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR