2-(Benzyloxy)-4-methyl-5-nitropyridine

Lieferant: Apollo Scientific

OR471283-1G
APOSOR471283-1GEA 214 EUR
APOSOR471283-1G
2-(Benzyloxy)-4-methyl-5-nitropyridine
2-(Benzyloxy)-4-methyl-5-nitropyridine
Formel: C₁₃H₁₂N₂O₃
Molecular Weight: 244,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909331
CAS-Nummer: 22754-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR