6-Chloro-2H-chromen-2-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR471287-250MG
APOSOR471287-250MGEA
81.2
EUR
APOSOR471287-250MG
6-Chloro-2H-chromen-2-one 95%
6-Chloro-2H-chromen-2-one
Formel:
C₉H₅ClO₂ Molecular Weight: 180,588 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01327472 CAS-Nummer: 2051-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...