4-(Aminomethyl)-3,5-dibromopyridine

Lieferant: Apollo Scientific

OR471290-1G
APOSOR471290-1GEA 916 EUR
APOSOR471290-1G
4-(Aminomethyl)-3,5-dibromopyridine
4-(Aminomethyl)-3,5-dibromopyridine
Formel: C₆H₆Br₂N₂
Molecular Weight: 265,933 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432803
CAS-Nummer: 1265895-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR