3-(Aminomethyl)-1,2,4-triazole

Lieferant: Apollo Scientific

OR471295-1G
APOSOR471295-1GEA 632 EUR
APOSOR471295-1G
3-(Aminomethyl)-1,2,4-triazole
3-(Aminomethyl)-1,2,4-triazole
Formel: C₃H₆N₄
Molecular Weight: 98.107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410878
CAS-Nummer: 58502-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR