4-(Aminomethyl)-3-bromopyridine

Lieferant: Apollo Scientific

OR471296-1G
APOSOR471296-1GEA 916 EUR
APOSOR471296-1G
4-(Aminomethyl)-3-bromopyridine
4-(Aminomethyl)-3-bromopyridine
Formel: C₆H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11100729
CAS-Nummer: 887581-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR