4-(Aminomethyl)-3-chloropyridine

Lieferant: Apollo Scientific

OR471297-1G
APOSOR471297-1GEA 916 EUR
APOSOR471297-1G
4-(Aminomethyl)-3-chloropyridine
4-(Aminomethyl)-3-chloropyridine
Formel: C₆H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11100728
CAS-Nummer: 870063-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR