[(5-Chloro-1H-indol-2-yl)methyl]amine

Lieferant: Apollo Scientific

Synonyme: (5-Chloro-1H-indol-2-yl)methanamine, 2-(Aminomethyl)-5-chloroindole

OR471299-1G
APOSOR471299-1GEA 288 EUR
APOSOR471299-1G
[(5-Chloro-1H-indol-2-yl)methyl]amine
[(5-Chloro-1H-indol-2-yl)methyl]amine
Formel: C₉H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02089399
CAS-Nummer: 21109-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR