N-Cbz-3-methyl-DL-valine methyl ester

Lieferant: Apollo Scientific

OR471304-1G
APOSOR471304-1GEA 601 EUR
APOSOR471304-1G
N-Cbz-3-methyl-DL-valine methyl ester
N-Cbz-3-methyl-DL-valine methyl ester
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279.332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11113181
CAS-Nummer: 141971-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR