2-Methylquinoline-5-boronic acid pinacol ester

Lieferant: Apollo Scientific

OR471311-1G OR471311-250MG
APOSOR471311-1GEA 369 EUR
APOSOR471311-1G APOSOR471311-250MG
2-Methylquinoline-5-boronic acid pinacol ester
2-Methylquinoline-5-boronic acid pinacol ester
Formel: C₁₆H₂₀BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22418301
CAS-Nummer: 876922-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR