(S)-2-Phenylpropanal

Lieferant: Apollo Scientific

OR471313-1G OR471313-250MG
APOSOR471313-1GEA 855 EUR
APOSOR471313-1G APOSOR471313-250MG
(S)-2-Phenylpropanal
(S)-2-Phenylpropanal
Formel: C₉H₁₀O
Lagertemperatur: Raumtemperatur
MDL: MFCD24673692
CAS-Nummer: 33530-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR