4-Chloro-6-(6-methoxy-3-pyridyl)quinazoline

Lieferant: Apollo Scientific

OR471328-1G
APOSOR471328-1GEA 1840 EUR
APOSOR471328-1G
4-Chloro-6-(6-methoxy-3-pyridyl)quinazoline
4-Chloro-6-(6-methoxy-3-pyridyl)quinazoline
Formel: C₁₄H₁₀ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD20482360
CAS-Nummer: 1005009-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR