3,7-Dibromo-2,6-dihydroxynaphthalene
Lieferant: Apollo Scientific
OR471332-1G
APOSOR471332-1GEA
672
EUR
APOSOR471332-1G
3,7-Dibromo-2,6-dihydroxynaphthalene
3,7-Dibromo-2,6-dihydroxynaphthalene
Formel:
C₁₀H₆Br₂O₂ Molecular Weight: 317,961 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28954613 CAS-Nummer: 1227743-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...