N-Boc-7-aminobenzofuran

Lieferant: Apollo Scientific

OR471333-1G
APOSOR471333-1GEA 774 EUR
APOSOR471333-1G
N-Boc-7-aminobenzofuran
N-Boc-7-aminobenzofuran
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233.263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954612
CAS-Nummer: 1934854-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR