N-Boc-2-phenylcyclopropylamine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (2-phenylcyclopropyl)carbamate

OR471350-1G OR471350-250MG
APOSOR471350-1GEA 409 EUR
APOSOR471350-1G APOSOR471350-250MG
N-Boc-2-phenylcyclopropylamine
N-Boc-2-phenylcyclopropylamine
Formel: C₁₄H₁₉NO₂
Molecular Weight: 233.306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21106315
CAS-Nummer: 92644-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR