4-Bromo-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

OR471353-1G
APOSOR471353-1GEA 1010 EUR
APOSOR471353-1G
4-Bromo-2-(hydroxymethyl)benzimidazole
4-Bromo-2-(hydroxymethyl)benzimidazole
Formel: C₈H₇BrN₂O
Molecular Weight: 227,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17267703
CAS-Nummer: 1248566-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR