4-Bromo-2-(hydroxymethyl)-6-methylbenzimidazole

Lieferant: Apollo Scientific

OR471354-1G
APOSOR471354-1GEA 1010 EUR
APOSOR471354-1G
4-Bromo-2-(hydroxymethyl)-6-methylbenzimidazole
4-Bromo-2-(hydroxymethyl)-6-methylbenzimidazole
Formel: C₉H₉BrN₂O
Molecular Weight: 241,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977369
CAS-Nummer: 1352519-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR