2-(Hydroxymethyl)-1H-imidazo[4,5-b]phenazine

Lieferant: Apollo Scientific

OR471355-1G
APOSOR471355-1GEA 804 EUR
APOSOR471355-1G
2-(Hydroxymethyl)-1H-imidazo[4,5-b]phenazine
2-(Hydroxymethyl)-1H-imidazo[4,5-b]phenazine
Formel: C₁₄H₁₀N₄O
Molecular Weight: 250,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977368
CAS-Nummer: 2006276-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR