2-(Hydroxymethyl)-5-(tert-butyl)benzimidazole
Lieferant: Apollo Scientific
OR471357-1G
APOSOR471357-1GEA
1010
EUR
APOSOR471357-1G
2-(Hydroxymethyl)-5-(tert-butyl)benzimidazole
2-(Hydroxymethyl)-5-(tert-butyl)benzimidazole
Formel:
C₁₂H₁₆N₂O Molecular Weight: 204,268 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977365 CAS-Nummer: 2006277-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...