6-Bromo-2-(hydroxymethyl)-4-methylbenzimidazole

Lieferant: Apollo Scientific

OR471358-1G
APOSOR471358-1GEA 1010 EUR
APOSOR471358-1G
6-Bromo-2-(hydroxymethyl)-4-methylbenzimidazole
6-Bromo-2-(hydroxymethyl)-4-methylbenzimidazole
Formel: C₉H₉BrN₂O
Molecular Weight: 241,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26912934
CAS-Nummer: 1565461-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR